Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
15-PGDH/HPGD Antibody (025), Alexa Fluor™ 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289840AF594
Description
15-PGDH/HPGD Monoclonal specifically detects 15-PGDH/HPGD in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
15-PGDH/HPGD | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
25 | |
ELISA, Sandwich ELISA Detection | |
15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human 15-PGDH/HPGD (NP_000851.2; Met1-Gln266). | |
0.1 mL | |
Cancer, Immunology, Innate Immunity | |
3248 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction