Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
15-PGDH/HPGD Antibody (101), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290625JF646
Description
15-PGDH/HPGD Monoclonal antibody specifically detects 15-PGDH/HPGD in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
15-PGDH/HPGD | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
101 | |
Western Blot, Immunoprecipitation | |
15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
Rabbit immunized with purified, recombinant Mouse 15-PGDH/HPGD (Uniprot#: Q8VCC1, Met1-Ser269) | |
0.1 mL | |
Cancer, Immunology, Innate Immunity | |
3248 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction