Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
15-PGDH/HPGD Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298291V
Description
15-PGDH/HPGD Polyclonal specifically detects 15-PGDH/HPGD in Mouse samples. It is validated for Western Blot, ELISA.Specifications
15-PGDH/HPGD | |
Polyclonal | |
Western Blot, ELISA | |
15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
Produced in rabbits immunized with purified, recombinant 15-PGDH/HPGD (Q8VCC1; Met 1-Ser 269). | |
0.1 mL | |
Cancer, Immunology, Innate Immunity | |
3248 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction