Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
15-PGDH/HPGD Rabbit anti-Human, Clone: 16, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | 15-PGDH/HPGD |
---|---|
Clone | 16 |
Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
15-PGDH/HPGD Monoclonal specifically detects 15-PGDH/HPGD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
15-PGDH/HPGD | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
Recombinant Human 15-PGDH/HPGD protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
16 | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Immunology, Innate Immunity | |
0.2 um filtered solution in PBS | |
3248 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title