Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-PGDH/HPGD Rabbit anti-Mouse, Clone: 101, Novus Biologicals™
SDP

Catalog No. NB024998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024998 50 μg
NB024997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB024998 Supplier Novus Biologicals Supplier No. NBP29062550UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

15-PGDH/HPGD Monoclonal specifically detects 15-PGDH/HPGD in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen 15-PGDH/HPGD
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 101
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias 15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1
Host Species Rabbit
Immunogen Recombinant Mouse 15-PGDH/HPGD protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3248
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.