Learn More
YSI™ DO Cap Membrane Kit
Catalog No. 15177560 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Includes:
Black Cap Membrane, 1.0 mil
Blue Cap Membrane, 2.0 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15-177-560 Black Cap Membrane, 1.0 mil
15-176-212 Blue Cap Membrane, 2.0 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-177-560 Supplier YSI™ Supplier No. 059880
Add to Cart
Add to Cart
DO Cap Membrane Kit for use with probes and electrodes.
  • DO Cap Membrane Kit
  • For use with sensors and probes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Instrument: 85, 550, and 556 MPS. Probes: 5239, 5905, 5010 and Pro Series Polarographic Sensors
Type DO Cap Membrane Kit
Flow Rate 12 in./sec.
Includes Black Cap Membrane, 1.0 mil
Material PTFE
Probe Type Oxygen/Temperature