Learn More
Fisherbrand™ 150 Handheld Homogenizer Motor Disposable Gernerator Probe Adapter
Shop All Fisherbrand Products
Click to view available options
Material:
Aluminum, Stainless Steel
Description
- Tip Adapter
- For motor with blue nose ring
- An Adapter is required to use Plastic Probes
- First, insert the smaller end of the Tip Adapter into the homogenizer as far as possible, turn clockwise, and release
- Next, insert an Tip Plastic Probe into the Tip Adapter as far as possible, turn clockwise, and release
- The homogenizer is now ready to operate. Tips can be disposed of after use.
Specifications
Specifications
| Product Type | Adapter |
| Material | Aluminum, Stainless Steel |
| Description | Not disposable |
| For Use With (Application) | Mounting Omni Tips™ to the Omni TH Homogenizer |