Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One 1536-well HiBase Polystyrene Microplates

Click to view available options
Color:
Black
White
Surface Treatment:
FLUOTRAC 200
LUMITRAC 200
Description
- Non-Pyrogenic
- Material: Polystyrene
- Free of detectable DNase, RNase, human DNA
- Traced way back to production through defined LOT number system
- Sample carryover and cross-contamination risk is minimized
- Total
Certifications
DIN ISO 9001

Specifications
Specifications
Format | Standard |
Lid | Without Lid |
Description | Plate design: HiBase |
Array | 32 x 48 |
Color | Black |
Material | Polystyrene |
No. of Wells | 1536 |
Sterility | Non-sterile |
Surface Treatment | FLUOTRAC 200 |
Well Shape | Flat Bottom (F-well) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction