Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trifluoromethanesulfonic Acid

Catalog No. 158534100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-853-4100G 100 g
15-853-450G 50 g
15-853-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 15-853-4100G Supplier Sigma Aldrich Supplier No. 158534100G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trifluoromethanesulfonic acid is the strongest monoprotic organic acid. It has been synthesized by the oxidation of bis(trifluoromethylthio)mercury with aqueous hydrogen peroxide. It undergoes complete dissociation in basic solvents such as dimethyl sulfoxide, dimethylacetamide and dimethylformamide. Its dissociation in non-aqueous solvents has been studied by conductometry. On mixing trifluoromethanesulfonic acid with HNO3, it forms nitronium trifluoromethane sulfonate, which is an excellent nitrating reagent.Trifluoromethanesulfonic acid may be used as an initiator for the cationic polymerization of styrene, hexamethylcyclotrisiloxane and L,L-dilactide.

Specifications

CAS 1493-13-6
Density 1.696 g/mL at 25°C
Boiling Point 162°C
Molecular Formula CHF3O3S
Refractive Index n20/D 1.327
Linear Formula CF3SO3H
MDL Number MFCD00007514
Quantity 100 g
Synonym TFMSA; Triflic acid
Molecular Weight (g/mol) 150.08 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.