Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

16-Hexadecanolide 97.0+%, TCI America™
SDP

Catalog No. H121925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H121925G 25 g
H12195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H121925G Supplier TCI America Supplier No. H121925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109-29-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00039668
InChI Key LOKPJYNMYCVCRM-UHFFFAOYSA-N
Synonym 16-hexadecanolide, hexadecanolide, cyclohexadecanolide, 16-hexadecanolactone, 1,16-hexadecanolide, dihydroambrettolide, juniperic acid lactone, 1,16-hexadecanolactone, hexadecanolactone, 16-hydroxyhexadecanoic acid lactone
PubChem CID 7984
IUPAC Name 1-oxacycloheptadecan-2-one
SMILES O=C1CCCCCCCCCCCCCCCO1

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 g
Formula Weight 254.41
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 16-Hexadecanolide
RTECSNumber RN9730000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.