Learn More
PIP 16 in. Black Polyblend Steel Toe and Shank Boot
Steel Toe Polyblend Waterproof Boot.Steel Shank. Pre-Formed Insole. ASTM F2413. Made in USA.
Supplier: PIP 383890/7
Description
- Less toe spring allows for more surface contact for improved channeling of water for better traction, slip resistance and comfort
- Steel Shank provides improved support and ladder stability
- Energy absorbing tread adds all-day comfort
- Tough outsole for good abrasion resistance. Flexible upper material stays supple in cold temperatures to make walking easier
- Midsole is designed to utilize additional material in the standard area of the shank providing increased support and comfort
- Lower profile 'kick off' ledge on back of boots for ease of removal
- Meets ASTM F3445-21 for Slip Resistance
- Meets ASTM F2413-18 M/I/C Impact and Compression
- Made in the USA
- Prop 65 Compliant
- Resistant to certain acids and alkalies, fats, oils hydrocarbons and other chemicals
Specifications
Black | |
7 | |
Unisex | |
Safety Boot |
PVC | |
Steel | |
Yes |