Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 17-AAG
GREENER_CHOICESDP

Catalog No. 15151 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-151 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-151 Supplier R&D Systems™ Supplier No. 1515/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective Hsp90 inhibitor

Inhibitor of heat shock protein 90 (Hsp90) chaperone activity, and an analog of geldanamycin (Cat. No. 1368). Subsequently inhibits the activity of oncogenic proteins such as p185erbB-2 (IC50 = 31 nM), N-ras, Ki-ras and c-Akt. Antitumor in vivo. Also protects neuroprogenitor cells against stress-induced apoptosis at low concentrations (10 nM) in vitro.

Chemical Identifiers

CAS 75747-14-7
Molecular Formula C31H43N3O8

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 585.7
Purity >98%
Molecular Weight (g/mol) 585.698
Chemical Name or Material 17-AAG
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.