Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
17-Beta-Estradiol Antibody (4S24 (BGN/06/8824)), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062280AF750
Description
17-Beta-Estradiol Monoclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.Specifications
17-Beta-Estradiol | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
NB100-62280 recognizes 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Estradiol 100% 17-beta Estradiol-BSA 90% Estriol <0.9% Testosterone <0.01% | |
Store at 4°C in the dark. |
ELISA | |
4S24 (BGN/06/8824) | |
ELISA | |
17 beta Oestradiol, 17-Beta-Estradiol, 17-beta-HSD | |
17-beta estradiol conjugated to bovine serum albumin at position 6. | |
Primary | |
All species | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction