Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17-Beta-Estradiol Antibody (4S24 (BGN/06/8824)) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$181.90 - $487.50

Specifications

Antigen 17-Beta-Estradiol
Clone 4S24 (BGN/06/8824)
Concentration 1 mg/mL
Dilution ELISA 1:100-1:2000
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017454
SDP
View Documents
Novus Biologicals
NB100622800.025MG
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10062280
SDP
View Documents
Novus Biologicals
NB10062280
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

17-Beta-Estradiol Monoclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.
Specifications

Specifications

17-Beta-Estradiol
1 mg/mL
ELISA
Unconjugated
RUO
17-beta estradiol conjugated to bovine serum albumin at position 6.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4S24 (BGN/06/8824)
ELISA 1:100-1:2000
Monoclonal
Mouse
All species
IgG1
Affinity Purified
NB100-62280 recognizes 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Estradiol 100% 17-beta Estradiol-BSA 90% Estriol <0.9% Testosterone <0.01%
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.