Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ ProVent™ 10000 Wrap-Around Gowns
SDP

Catalog No. 17988497e Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-497E 2X-Large
17-988-497G 4X-Large
17-988-497A Small
17-988-497C Large
17-988-497B Medium
17-988-497D X-Large
17-988-497F 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-988-497E Supplier Kappler™ Supplier No. PPN101BU2X
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

ProVent 10,000 wrap-around gowns feature a patented microporous film composite fabric that offers a wide range of barrier protection.

  • ProVent 10,000 is a patented microporous film composite fabric that offers a wide range of barrier protection against blood-borne pathogens, body fluids, and viral threats
  • It passes both ASTM F1670 blood penetration and ASTM F1671 viral penetration tests
  • High moisture vapor transmission rate (MVTR) provides a more comfortable garment ideal for biohazard work applications

Compliance

Meets ASTM™ F1670 and ASTM F1671 standards.

Specifications

No. of Pockets None
For Use With (Application) Biohazard Cleanup, Healthcare, Maintenance
Size 2X-Large
Gender Unisex
Product Type General Purpose Gown
Material Composite Fabric
Color Blue
Closure Type Hook and Loop
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model MN101BU2X
Seam Type Ultrasonic
Sleeve Style Long Sleeve
Certifications/Compliance ASTM F1670, ASTM F1671
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.