Learn More
Pyramex™ Venture 3™ Clear Anti-Fog Lens with Black Temples
Soft dual-injected rubber temple tips combined with fully adjustable straight back temples reinforce high comfort and fit
Supplier:  Pyramex™ SB5710DT
 View more versions of this product
Catalog No. 19-817-535
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Soft and adjustable ventilated nose piece 
  • Top of frame ventilated allowing air to circulate
  • 9.5 base curve lens provides excellent side protection
  • Scratch-resistant polycarbonate lens provide 99% UVA/B/C protection
Specifications
Specifications
Scratch-resistant
Black
Polycarbonate
Clear
Fully adjustable straight back temples
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.