Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell X-Tra™ Traction Shoe Covers
GREENER_CHOICE

Catalog No. 190202502 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium/Large
X-Large/2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-2502 X-Large/2X-Large
19-020-2501 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-020-2502 Supplier Ansell Supplier No. 69254
Add to Cart
Add to Cart

Shoe covers made with SMS fabric feature adhesive traction strips on the bottom for stability.

  • Light blue, three-layer SMS fabric
  • Fully elasticized
  • Adhesive strips for traction on bottom
TRUSTED_SUSTAINABILITY

Specifications

Size X-Large/2X-Large
Material SMS
Coating Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Traction Strips
Model 69254
Includes Traction
For Use With (Application) Laboratories, Healthcare
Packaging Quantity 240/Cs.
Product Type Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.