Learn More
Ansell KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
Catalog No. 19057450C Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
Hood Type:
Attached
Elastic
Ankle Style:
Attached Boots
Elastic
Seam Type:
Taped
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hood Type Ankle Style Seam Type
19-057-450C 2X-Large Attached Attached Boots Taped
19-057-449B X-Large Elastic Elastic Taped
19-057-450D 3X-Large Attached Attached Boots Taped
19-057-449C 2X-Large Elastic Elastic Taped
19-057-449A Large Attached Elastic Taped
19-057-450B X-Large Attached Attached Boots Taped
19-057-449E 4X-Large Attached Elastic Taped
19-057-449D 3X-Large Elastic Elastic Taped
19-057-450E 4X-Large Attached Attached Boots Taped
19-057-450A Large Attached Attached Boots Taped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-057-450C Supplier Ansell Supplier No. 45665
Add to Cart
Add to Cart
This item is not returnable. View return policy
For chemical permeation and jet liquid protection
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
  • Zipper front with taped storm flap
  • Interior has comfortable, cloth-like feel against the skin
  • Reflex™ Design provides 12% more chest room and 6% longer body length
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Saranex 23-P Fabric
Color White
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper with Storm Flap
Seam Type Taped
Resistance Chemicals
Description Hooded (respirator-fit), zipper front with storm flap, elastic wrist and ankle, exterior lamination; Saranex™ 23-P Film Laminated to Durable Fabric
For Use With (Application) Chemical handling or mixing or cleanup, hazardous material handling, hazardous waste cleanup, industrial hazmat teams, petrochemicals, utilities and for chemical permeation and jet liquid protection
Weight (English) 1.42 lb.
Weight (Metric) 0.64 kg