Learn More
3M™ SecureFit™ 3700 Series Safety Glasses
Shop All 3M Products
Click to view available options
Lens Tint:
Blue
IR/UV 5.0, Gray
Orange
Shade 1.7, Gray
Description
- Pressure Diffusion Temple (PDT) Technology helps diffuse pressure over the ear for a secure and comfortable fit
- Adjustable Ratchet Temples provide five adjustment points on the temple to help create a secure and personalized fit
- A soft touch nosepiece helps provide longwearing comfort
- Predominantly used with other application specific PPE
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | ANSI Z87.1-2015 |
| Description | Model: SF3717AS-BLK |
| Lens Tint | Shade 1.7, Gray |
| Product Type | Safety Glasses |
| For Use With (Application) | Pharmaceuticals, Oil and Gas, Machine Operations, Law Enforcement, Chemical Handling, Food and Beverage Manufacturing, Riveting, Assembly, Construction, Sub Applications, Pouring and Casting, Chemicals, General Manufacturing, Sawing, Mining, Facility, Maintenance, UV Protection, Primary Metals, Remediation, Chiseling, Aviation, Machining, Automotive, Demolition, Transportation, Masonary, Drilling, Grinding, Food and Beverage, Power Fastening |
| Frame Color | Clear |
| No. per Case | 10/Cs. |
| Temple Style | 3M Pressure Diffusion Temple Technology (PDT) |
| Temple Color | Black |
| Show More |