Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Bandage Spray

Catalog No. 19090652 Shop All Medique Products
Change view
Click to view available options
Includes:
3 oz. can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-090-652 3 oz. can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-090-652 Supplier Medique Supplier No. 45017
Add to Cart
Add to Cart
This item is not returnable. View return policy

First aid aerosol spray for minor cuts, burns, skin irritations, insect bites and poison ivy.

  • Provides protective coating
  • Water soluble

Specifications

Product Type Bandage Aerosol Spray
For Use With (Application) Provides protective coating for minor cuts, burns, skin irritations, insect bites, poison ivy
Format Water Soluble
Includes 3 oz. can
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.