Learn More
Pyramex™ Ever-Lite™ Safety Eyewear
Catalog No. 19155846 Shop All Pyramex Safety Products Products
Click to view available options
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Dark Gray
Gray
Indoor/Outdoor, Mirror
Photochromic
Lens Coating:
Anti-fog
Uncoated
This item is not returnable. View return policy
This item is not returnable. View return policy
Lightweight spectacle (22 grams) with a soft nosepiece for snug fit, semi-frame, and slim co-injected temples
  • Offers a clear panoramic view without obstruction
  • 8 base wraparound lens provides full front side protection
  • Scratch-resistant, polycarbonate lens provide 99.9% of UV protection
  • H2MAX creates a high performing, superior anti-fog coating that is durable enough to withstand repeated cleaning without losing adhesion
  • H2MAX ensures maximum clarity in the most extreme temperature and humidity conditions, and it also adds a layer to the lens that is abrasion, chemical, and UV-resistant
  • Specifications
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Gray
    Frame Color Black
    Weight (Metric) 22 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.