Learn More
Kimtech™ M7 Face Veils
GREENER_CHOICE
Catalog No. 19165385 Shop All Ansell Products
Change view
Click to view available options
Fastening Type:
Headband
Snaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type
19-165-385 Snaps
19-160-0417 Headband
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-165-385 Supplier Kimtech™ Supplier No. 62751
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable face veils with knitted headbands or snaps are constructed of low-lint polyethylene film. Covered nose piece provides increased comfort. Suitable for ISO Class 7 or higher cleanroom environments.
  • Constructed of low-lint polyethylene film
  • Covered nose piece to provide increased comfort
  • Ultra sonic bond for higher product integrity
  • Latex and silicone free
  • Bacterial and particle filtration to avoid contamination
  • Suitable for use in ISO 7 or higher cleanrooms
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Veil
Color White
Fastening Type Snaps
Material Polyester
Size Standard
Sterility Non-sterile
Style Veil
Cleanroom Class Class 10,000 (ISO 7)
Disposable Single-use
Includes Stainless Steel Snaps
Latex Free Yes
Length (English) 15 in.
Length (Metric) 38.1 cm
Model 62751
Packaging Type Double-bagged
Series Kimtech™ M7
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.