Learn More
STERIS LpH™ III se Phenolic Disinfectant
Shop All STERIS Corporation ProductsDescription
This STERIS™ LpH™ III se Disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the LpH™ III Disinfectants kill a broad spectrum of common bacteria and fungi. The acidic LpH™ III Disinfectant formulation can be used alone or in rotation with the Vesphene™ III Disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS™ Technical Services team for individualized application and validation assistance.
How LpH™ III se Disinfectant Works
STERIS'™ LpH™ III se Phenolic Disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
Why LpH™ III se Disinfectant?
- Efficacy
- Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
- Productivity
- Formulation includes surfactants for surface cleaning while disinfecting.
- Flexibility
- Ideal for use in a contamination control rotation.
- Reliability
- Complete lot traceability.
- Assurance
- Each lot documented by Certificate of Analysis.
Specifications
Specifications
Product Type | Phenolic Disinfectant |
Form | Concentrated Liquid |
pH | 2.1 |
Sterility | Non-sterile |
Concentration | 0.8% v/v (1:128) |
Container Type | Pail |
For Use With (Application) | Cleanroom, Controlled Environments |
Volume (English) | 5 gal. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.