Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorLite™ Full Nitrile Coated Cotton Gloves with Textured Finish

Catalog No. 19816816 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-816 Large
19-123-379A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-816-816 Supplier PIP™ Supplier No. 563171/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Armor™ series gloves feature a unique combination of low-coating deposit weight and high abrasion resistance. Gloves provide the wear of heavier, bulkier nitrile coated gloves without sacrificing flexibility and tactile sensitivity.

  • ActiFresh™ treated to reduce germs and odors
  • Thinly coated with blue nitrile on cotton interlock liner
  • Gloves are comfortable, flexible, and increase dexterity
  • Textured coating is resistant to grease and oils for enhanced grip capabilities
  • Effective, affordable protection from physical hazards such as cuts, punctures, snags and abrasion
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Made in ISO 9001:2000 / ISO 14001:2000 facilities
  • Launderable

Specifications

Coated Area Palm, Fingers
Coating Material Nitrile
Color Blue
Exterior Finish Fully Textured
Hand Specific
Product Type Dip-coated Gloves
Size Large
For Use With (Application) General Handling, Assembly, Fabrication, Inspection, Maintenance, and Shipping/Receiving
Glove Cuff Color Natural
Glove Cuff Style Knit
Liner Material Cotton
Series 56-3171
Length (English) 10.8 in.
Length (Metric) 27.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.