Learn More
STERIS Cage-Klenz 180 Alkaline Detergent
Shop All STERIS Corporation Products
Click to view available options
Size:
5 gal.
30 gal.
55 gal.
250 gal.
Description
- Cage-Klenz 180 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in research animal caging.
- Cage-Klenz 180 Detergent is a liquid, phosphate-free detergent for use in high pressure spray animal cage washing equipment. It is formulated with corrosion inhibitors for enhanced materials compatibility.
- Efficacy - Effectively cleans animal fats, oils and other organic cage residues in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - phosphate-free formulation
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Specifications
Product Type | Detergent |
Color-coded | No |
Form | Clear Liquid |
Scent | Unscented |
Sterility | Non-sterile |
Size | 30 gal. |