Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ Chill-Its™ 6615 High-Performance Dew Rag
Shop All Ergodyne Products
Click to view available options
Color:
Black
Gray, Black, White
Lime
Realtree Xtra™
Description
- Hi Cool™ moisture management performance fabric
- Comfortable to wear under hats, helmets, or alone
- Absorbent terry headband
- Elastic band for a great fit
- Available in a variety of patterns and colors like black, lime, skulls, and Realtree Xtra™
Specifications
Specifications
Material | Hi Cool™ Moisture Management Performance Fabric |
Product Type | Dew Rag |
Color | Gray, Black, White |
Band Type | Elastic |
For Use With (Application) | Construction, Maintenance, Landscaping |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction