Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell TouchNTuff™ 93-300 Clean Nitrile Gloves
BIOPHARMAGREENER_CHOICE

Catalog No. 19823525 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-525 X-Large
19-823-522 Small
19-823-523 Medium
19-823-524 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-823-525 Supplier Ansell Supplier No. 556082
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-sterile, clean nitrile gloves are compatible with Class 100 (ISO 5) controlled environments. Disposable gloves offer three times the puncture and tear resistance of standard latex gloves.

  • Clean nitrile glove
  • Compatible with Class 100 (ISO 5)/Grade A Cleanroom Environments
  • Chemical splash protection
  • Resists punctures and tears
  • Silicone-free for product protection
  • Recommended for Biotechnology Manufacturing industries
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6319, FDA 21 CFR 177.2600, ISO 13485, ISO 14001
Certifications/Compliance (Europe) Category III, EAC TP TC 019:2011, EN 420:2003 + A1:2009, EN ISO 374-1:2016, EN ISO 374-5:2016, EN421:2010
Cleanroom Class Class 100 (ISO 5)
Color Green
Exterior Finish Partially Textured
Finger Thickness (English) 6.3 mil
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.1 mil
Palm Thickness (Metric) 0.13 mm
Size X-Large
Sterility Non-sterile
Thickness (English) 6.3 mil
Thickness (Metric) 0.16 mm
Manufacturing/QMS Audit Standards ISO 9001:2015
Model 556082
Series TouchNTuff™ 93-300
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.