Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one 95.0+%, TCI America™
SDP

Catalog No. H1525100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
20 mg
100 mg

Chemical Identifiers

CAS 1217503-60-0
Molecular Formula C22H24O7
Molecular Weight (g/mol) 400.427
InChI Key SKZNJIJSGSQNJQ-CDJQDVQCSA-N
Synonym GO-Y078
PubChem CID 46231908
IUPAC Name (1E,4E)-1-(4-hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)penta-1,4-dien-3-one
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)C=CC2=CC(=C(C(=C2)OC)OC)OC

Specifications

Quantity 100 mg
Formula Weight 400.43
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.