Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Heptadecafluoro-1-decanol 98.0+%, TCI America™
SDP

Catalog No. H084525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 678-39-7
Molecular Formula C10H5F17O
Molecular Weight (g/mol) 464.122
MDL Number MFCD00039544
InChI Key JJUBFBTUBACDHW-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluoro-1-decanol, 1h,1h,2h,2h-perfluorodecanol, 2-perfluoro-n-octyl ethanol, 1,1,2,2-tetrahydroperfluoro-1-decanol, 2-perfluorooctyl ethanol, 8:2 fluorotelomer alcohol, 1h,1h,2h,2h-perfluorodecan-1-ol, 8:2 ftoh, unii-um7l66ofe1, 1-decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro
PubChem CID 69619
ChEBI CHEBI:63919
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol
SMILES C(CO)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 25 g
Formula Weight 464.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H,2H,2H-Heptadecafluoro-1-decanol
EINECSNumber (2)-2402
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.