Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H-Perfluoro-1-octanol, 98%

Catalog No. p-3779388
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 307-30-2 | C8H3F15O | 400.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-30-2
Molecular Formula C8H3F15O
Molecular Weight (g/mol) 400.08
MDL Number MFCD00004675
InChI Key PJDOLCGOTSNFJM-UHFFFAOYSA-N
Synonym 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, pentadecafluorooctyl alcohol, unii-9x5w7fq62r, 1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, perfluorooctanol
PubChem CID 67543
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol
SMILES C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color White
Boiling Point 100°C to 102°C (100.0 hPa)
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)6CH2OH
Quantity 5 g
Beilstein 01, IV, 1766
Solubility Information Solubility in water: insoluble
Formula Weight 400.08
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 1H, 1H-Perfluoro-1-octanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 206-197-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.