Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-indazole-3-carboxylic acid, Thermo Scientific™

Catalog No. MO07468DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07468DE 5 g
MO07468DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO07468DE Supplier Thermo Scientific Chemicals Supplier No. MO07468DE

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 4498-67-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
InChI Key BHXVYTQDWMQVBI-UHFFFAOYSA-N
Synonym indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
PubChem CID 78250
IUPAC Name 1H-indazole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=NN2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1H-Indazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.