Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazole-5-boronic acid, 97%, Thermo Scientific™

Catalog No. ac430350010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 338454-14-1
Molecular Formula C7H7BN2O2
Molecular Weight (g/mol) 161.96
MDL Number MFCD06739151
InChI Key CLVPGJWAMIADSY-UHFFFAOYSA-N
Synonym 1h-indazole-5-boronic acid, 1h-indazol-5-yl boronic acid, indazole-5-boronic acid, 1h-indazol-5-yl-5-boronic acid, boronic acid, b-1h-indazol-5-yl, boronic acid, b-2h-indazol-5-yl, b-1h-indazol-5-yl-boronic acid, 5-borono-1h-indazole, 5-indazolylboronic acid, pubchem23544
PubChem CID 17750504
IUPAC Name 1H-indazol-5-ylboronic acid
SMILES OB(O)C1=CC=C2NN=CC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazole-5-boronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.