Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazole, 99%

Catalog No. AAA1166514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1166514 25 g
AAA1166503 1 g
AAA1166506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1166514 Supplier Thermo Scientific Chemicals Supplier No. A1166514
Only null left
Add to Cart

CAS: 271-44-3 | C7H6N2 | 118.139 g/mol

1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

Solubility
Soluble in hot water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-44-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00005691
InChI Key BAXOFTOLAUCFNW-UHFFFAOYSA-N
Synonym indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
PubChem CID 9221
ChEBI CHEBI:36669
IUPAC Name 1H-indazole
SMILES C1=CC=C2C(=C1)C=NN2

Specifications

Melting Point 146°C to 149°C
Boiling Point 270°C
Quantity 25 g
Beilstein 109676
Merck Index 14,4936
Solubility Information Soluble in hot water.
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 1H-Indazole
EINECSNumber 205-978-4
RTECSNumber NK7745000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.