Learn More
1H-Indene, 90+%, stab. with 0.01% 4-tert-butylcatechol
Description
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.
Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.
Notes
Store at 4°C. Store in dark. Incompatible with oxidizing agent, light, heat.
Specifications
Specifications
| Melting Point | ∼-2°C |
| Density | 0.996 |
| Boiling Point | 181°C to 182°C |
| Flash Point | 58°C (136°F) |
| Odor | Aromatic |
| Refractive Index | 1.57 |
| Quantity | 250 mL |
| UN Number | UN3295 |
| Beilstein | 635873 |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H226-H335-H373 |
| missing translation for 'healthHazard2' | GHS H Statement H304-H226-H332 May be fatal if swallowed and enters airways. Flammable liquid and vapor. Harmful if inhaled. |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 202-393-6 |
| RTECSNumber | NK8225000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.