Learn More
1H-Indene, 97%
Description
1H-Indene is used in the synthesis of new C60 derivative, indene-C60 bisadduct1. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2. It is also used in the synthesis of istatins and in the production of indene/cumarone thermoplastic resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1H-Indene is used in the synthesis of new C60 derivative, indene-C60 bisadduct1. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2. It is also used in the synthesis of istatins and in the production of indene/cumarone thermoplastic resins.
Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents Recommended storage temperature is 2 - 8°C. It is sensitive to light.
Specifications
Specifications
| Melting Point | ∼-2°C |
| Density | 0.996 |
| Boiling Point | 181°C to 182°C |
| Flash Point | 58°C (136°F) |
| Odor | Aromatic |
| Refractive Index | 1.574 |
| Quantity | 5 g |
| UN Number | UN3295 |
| Beilstein | 635873 |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H335-H373 |
| Hazard Statement | GHS H Statement H304-H226-H332 May be fatal if swallowed and enters airways. Flammable liquid and vapor. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 202-393-6 |
| RTECSNumber | NK8225000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.