Learn More
1H-Indole-3-acetic acid, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• 1H-Indole-3-acetic acid is a plant growth hormone that belongs to the auxin class. It is also considered to be an inducer of plant cell elongation and division that can cause uncontrolled growth
Applications
• This compound is a monocarboxylic acid; an acetic acid in which one of the methyl hydrogens has been replaced by a 1H-indol-3-yl group.
• Beyond its role as a plant hormone, this compound regulates tissue differentiation, apical dominance, and responses to light, gravity, and pathogens
• It can be used as a signaling molecule necessary for development of plant organs and coordination of growth
 
            Specifications
Specifications
| CAS Min % | 99.0 | 
| CAS Max % | 100.0 | 
| Melting Point | 167.0°C | 
| Color | Colorless to Yellow | 
| Flash Point | 171°C | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 99% min. (HPLC) | 
| Packaging | Glass bottle | 
| Quantity | 25 g | 
| Beilstein | 22, 66 | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Warning | 
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. | 
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee | 
| EINECSNumber | 201-748-2 | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                