Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indole-4-carbaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC28704DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 1074-86-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD01632221
InChI Key JFDDFGLNZWNJTK-UHFFFAOYSA-N
Synonym indole-4-carboxaldehyde, 4-formylindole, 1h-indole-4-carboxaldehyde, indole-4-carbaldehyde, 4-indole-carboxaldehyde, 4-indolecarbaldehyde, 4-formyl-indole, 4-formyl-1h-indole, 1h-indol-4-carbaldehyde, indole-4-aldehyde
PubChem CID 333703
IUPAC Name 1H-indole-4-carbaldehyde
SMILES O=CC1=C2C=CNC2=CC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 145.16
Percent Purity 95%
Chemical Name or Material 1H-indole-4-carbaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.