Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Pyrazole-3-boronic acid pinacol ester, 95%

Catalog No. p-7030083
Click to view available options
Quantity:
1 g
5 g

CAS: 844501-71-9 | C9H15BN2O2 | 194.041 g/mol

1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844501-71-9
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.041
MDL Number MFCD07368044
InChI Key KWLOIDOKWUESNM-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-3-boronic acid pinacol ester, pyrazole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-pyrazole, 3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-5-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxabolone-pyrrazole, 1h-pyrazol-3-yl-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolone-pyrrazole
PubChem CID 21931539
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2

Specifications

Melting Point 105°C to 108°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 194.04
Percent Purity 95%
Chemical Name or Material 1H-Pyrazole-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.