Learn More
Thermo Scientific™ 1M Triethylammonium Bicarbonate (TEAB) Buffer, 50 mL PROMO
Description
Triethylamonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.
Specifications
Specifications
| Product Type | Triethylammonium Bicarbonate (TEAB) |
| Product Line | TMT |
| Content And Storage | Store protected from direct sunlight in a dry, cool, and well-ventilated area. |
| Starting Material | Cell Lysate |
| Workflow Step | Protein Labeling |
| Quantity | 50 mL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.