Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-1-(2,5-Dimethoxy-2,5-dihydrofuran-2-yl)ethanol, 98%, Thermo Scientific™

Catalog No. AC339730010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 316186-17-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD01863302
InChI Key OHXZDCYIALCSNL-JECWYVHBSA-N
Synonym 1r-1-2,5-dimethoxy-2h-furan-5-yl ethanol, 1r-1-2,5-dimethoxy-5h-furan-2-yl ethanol, 2-furanmethanol,2,5-dihydro-2,5-dimethoxy-a-methyl-, ar, 1r-1-2,5-dimethoxy-2,5-dihydrofuran-2-yl ethanol, 98
PubChem CID 2733897
IUPAC Name (1R)-1-(2,5-dimethoxy-2H-furan-5-yl)ethanol
SMILES CC(C1(C=CC(O1)OC)OC)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4555 to 1.4575 (20°C, 589nm)
Quantity 1 g
Specific Rotation +9° to +11° (20°C, 589nm) (c=1, ethanol)
Formula Weight 174.2
Percent Purity ≥97.5%
Physical Form Viscous Liquid
Chemical Name or Material (1R)-1-(2, 5-Dimethoxy-2, 5-dihydrofuran-2-yl)ethanol, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.