Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%

Catalog No. p-7034606
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 46022-05-3 | C8H12O4 | 172.18 g/mol

Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.

Solubility
Soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 46022-05-3
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00062973
InChI Key QSAWQNUELGIYBC-PHDIDXHHSA-N
Synonym 1r,2r-cyclohexane-1,2-dicarboxylic acid, trans-1,2-cyclohexanedicarboxylic acid, trans-cyclohexane-1,2-dicarboxylic acid, 1r,2r---1,2-cyclohexanedicarboxylic acid, 1r,2r-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2r-rel, 1,2-cyclohexanedicarboxylic acid, 1r,2r, 1r.2r-cyclohexane-1,2-dicarboxylicacid, 1,2-cyclohexanedicarboxylic acid, trans, pubchem19603
PubChem CID 720895
IUPAC Name (1R,2R)-cyclohexane-1,2-dicarboxylic acid
SMILES C1CCC(C(C1)C(=O)O)C(=O)O

Specifications

Quantity 250 mg
Solubility Information Soluble in acetone.
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material (1R,2R)-Cyclohexane-1,2-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.