Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine, 98+%

Catalog No. p-7028662
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 144222-34-4 | C21H23N2O2S | 367.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144222-34-4
Molecular Formula C21H23N2O2S
Molecular Weight (g/mol) 367.49
MDL Number MFCD02093428
InChI Key UOPFIWYXBIHPIP-NHCUHLMSSA-O
Synonym 1r,2r---n-4-toluenesulfonyl-1,2-diphenylethylenediamine, 1r,2r---n-p-tosyl-1,2-diphenylethylenediamine, r,r-tsdpen, r,r-n-p-toluenesulfonyl-1,2-diphenylethylenediamine, 1r,2r-n-p-tosyl-1,2-diphenylethylenediamine, n-1r,2r-2-amino-1,2-diphenylethyl-4-methylbenzenesulfonamide, r,r-n-2-amino-1,2-diphenylethyl-p-toluenesulfonamide, 1r,2r---n-p-tosyl-1,2-diphenylethylene diamine, 1r, 2r---n-4-toluene sulfonyl 1,2-diphenyl-1,2-ethane diamine, benzenesulfonamide, n-1r,2r-2-amino-1,2-diphenylethyl-4-methyl
PubChem CID 2734565
IUPAC Name N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 127°C to 132°C
Quantity 1 g
Optical Rotation −31° (c=1 in Chloroform)
Formula Weight 366.48
Percent Purity ≥98%
Chemical Name or Material (1R,2R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.