Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,3S)-3-Aminocyclopentanecarboxylic acid, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
71830-08-5
C6H11NO2
129.16
MLLSSTJTARJLHK-UHNVWZDZSA-N
1r,3s-3-aminocyclopentanecarboxylic acid,1r,3s-3-aminocyclopentane-1-carboxylic acid,--1r,3s-3-aminocyclopentanecarboxylic acid,cis-3-aminocyclopentanecarboxylic acid,cyclopentanecarboxylic acid, 3-amino-, 1r,3s,1r,3s---3-aminocyclopentanecarboxylic acid,cis-3-amino-cyclopentanecarboxylic acid,1r,3s-3-amino-cyclopentanecarboxylic acid,pubchem18731,1r,3s---3-amino-cyclopentanecarboxylic acid
1502035
(1R,3S)-3-aminocyclopentane-1-carboxylic acid
C1CC(CC1C(=O)O)N
71830-08-5
C6H11NO2
129.16
MLLSSTJTARJLHK-UHNVWZDZSA-N
1r,3s-3-aminocyclopentanecarboxylic acid,1r,3s-3-aminocyclopentane-1-carboxylic acid,--1r,3s-3-aminocyclopentanecarboxylic acid,cis-3-aminocyclopentanecarboxylic acid,cyclopentanecarboxylic acid, 3-amino-, 1r,3s,1r,3s---3-aminocyclopentanecarboxylic acid,cis-3-amino-cyclopentanecarboxylic acid,1r,3s-3-amino-cyclopentanecarboxylic acid,pubchem18731,1r,3s---3-amino-cyclopentanecarboxylic acid
1502035
(1R,3S)-3-aminocyclopentane-1-carboxylic acid
C1CC(CC1C(=O)O)N
Enantiomeric Excess 97.5% (HPLC) min.
Melting Point 172.1°C
Quantity 1g
Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
Assay Percent Range 95%
Formula Weight 129.16
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material (1R, 3S)-3-Aminocyclopentanecarboxylic acid
Show More Show Less