Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1R,3S)-(+)-Camphoric Acid

Catalog No. 503119131 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
5 g
25 g

Catalog No. 50-311-9131

Supplier: Sigma Aldrich C409100G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1R,3S)-(+)-Camphoric Acid

Specifications

CAS 124-83-4
Melting Point 183°C to 186°C
Molecular Formula C10H16O4
Linear Formula C10H16O4
MDL Number MFCD00001375
Quantity 5 g
Synonym (+)-Camphoric acid; (1R,3S)-1,2,2-Trimethyl-1,3-cyclopentanedicarboxylic acid
Molecular Weight (g/mol) 200.23 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.