Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-(-)-Menthyl acetate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. p-7187624 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 88-72-2
Density 0.92 g/mL (at 25°C)
Flash Point 77°C
Physical Form Neat
Linear Formula C12H22O2
MDL Number MFCD00001482
Boiling Point 229°C to 230°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 198.30
Percent Purity ≥99.0% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.