Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-(+)-alpha-Pinene, 98%, 80% ee

Catalog No. AC131262500
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Glass bottle

CAS: 7785-70-8 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-70-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001346
InChI Key GRWFGVWFFZKLTI-RKDXNWHRSA-N
Synonym +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene
PubChem CID 82227
ChEBI CHEBI:28261
SMILES CC1=CC[C@@H]2C[C@H]1C2(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -62.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 155.0°C
Flash Point 33°C
Enantiomeric Excess 75% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 250 g
Beilstein 05, 146
Fieser 15,201
Specific Gravity 0.86
Merck Index 14, 7445
Specific Rotation Condition +43.50° (20°C neat)
Specific Rotation + 43.50
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform,diethyl ether
IUPAC Name (1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
Formula Weight 136.24
Percent Purity 98%,80% ee
Physical Form Liquid
Chemical Name or Material (1R)-(+)-α-Pinene, 98% (80+% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
EINECSNumber 232-087-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.