Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
28143-91-1
C9H13NO2
167.21
MFCD00004503
JUCGVCVPNPBJIG-IUCAKERBSA-N
1s,2s-+-2-amino-1-phenyl-1,3-propanediol,1s,2s-2-amino-1-phenylpropane-1,3-diol,l-+-threo-2-amino-1-phenyl-1,3-propanediol,1s,2s-2-amino-1-phenyl-1,3-propanediol,1s,2s-2-amino-1-phenyl-propane-1,3-diol,1s, 2s-+-2-amino-1-phenyl-1,3-propanediol,1,3-propanediol, 2-amino-1-phenyl-, 1s,2s,pubchem6415,s r*,r*-2-amino-1-phenylpropane-1,3-diol,s,s-2-amino-1-phenyl-1,3-propanediol
119881
(1S,2S)-2-amino-1-phenylpropane-1,3-diol
C1=CC=C(C=C1)C(C(CO)N)O
28143-91-1
C9H13NO2
167.21
MFCD00004503
JUCGVCVPNPBJIG-IUCAKERBSA-N
1s,2s-+-2-amino-1-phenyl-1,3-propanediol,1s,2s-2-amino-1-phenylpropane-1,3-diol,l-+-threo-2-amino-1-phenyl-1,3-propanediol,1s,2s-2-amino-1-phenyl-1,3-propanediol,1s,2s-2-amino-1-phenyl-propane-1,3-diol,1s, 2s-+-2-amino-1-phenyl-1,3-propanediol,1,3-propanediol, 2-amino-1-phenyl-, 1s,2s,pubchem6415,s r*,r*-2-amino-1-phenylpropane-1,3-diol,s,s-2-amino-1-phenyl-1,3-propanediol
119881
(1S,2S)-2-amino-1-phenylpropane-1,3-diol
C1=CC=C(C=C1)C(C(CO)N)O
Specific Rotation + 35.00
Infrared Spectrum Authentic
Melting Point 110.0°C to 114.0°C
Color White to Yellow
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH(OH)CH(NH2)CH2OH
Formula Weight 167.21
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material (1S, 2S)-(+)-2-Amino-1-phenyl-1, 3-propanediol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 248-867-6

TSCA : TSCA