Learn More
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
Description
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Synonym | 1s,2s-2-aminocyclohexanecarboxylic acid, 1s,2s-2-aminocyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 2-amino-, 1s,2s, 1s,2s-2-aminocyclohexanecarboxylicacid, 1s,2s-2-amino-cyclohexanecarboxylic acid, 2beta-amino-1alpha-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid,2-amino-, 1s,2s, 24716-92-5 1s,2s-2-aminocyclohexane-1-carboxylic acid |
| Solubility Information | Soluble in water. |
| Quantity | 5 g |
| Optical Rotation | +77° (c=1 in Water) |
| Formula Weight | 143.19 |
| Percent Purity | 97% |
| Chemical Name or Material | (1S,2S)-2-Aminocyclohexanecarboxylic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.