Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee

Catalog No. AC361840010
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle

CAS: 161660-94-2 | C11H18NO4 | 228.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161660-94-2
Molecular Formula C11H18NO4
Molecular Weight (g/mol) 228.27
MDL Number MFCD01320858,MFCD01320857
InChI Key RNJQBGXOSAQQDG-SFYZADRCSA-M
Synonym 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
PubChem CID 1512526
SMILES CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O

Specifications

Melting Point 96.1°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
IUPAC Name (1R,3S)-3-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Quantity 1 g
Formula Weight 229.27
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (1S, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.