Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 1S,3R-RSL3
GREENER_CHOICESDP

Catalog No. 611850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-185-0 50 mg
61-181-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-185-0 Supplier Tocris Bioscience™ Supplier No. 6118/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Induces ferroptosis; inhibits glutathione peroxidase 4 (GPX4)

1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.

Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.

Specifications

Chemical Name or Material (1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate
CAS 1219810-16-8
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C23H21ClN2O5
Formula Weight Observed MW: 440.88
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.